Learn More
AQUABILITI WFI 1100L BAG USP GRADE
Catalog No. NC3947808
Encompass_Preferred
AQUAPure ULTRA is a high-purity water for injection (WFI) quality water, meeting USP grade standards for critical laboratory and pharmaceutical applications. This high-quality water is engineered to provide low impurity profiles, ensuring reliability for sensitive experimental and manufacturing procedures.
  • Meets USP grade requirements for water for injection
  • Low conductivity of 0.06 µS/cm
  • Total organic carbon (TOC) content of 50 ppb
  • Endotoxin levels controlled at 0.03 EU/mL
  • Supplied in a 1100L bag configuration
Catalog No. NC3947808 Supplier AQUABILITI Supplier No. ULTRA110001
AQUAPure ULTRA is a high-purity water for injection (WFI) quality water, meeting USP grade standards for critical laboratory and pharmaceutical applications. This high-quality water is engineered to provide low impurity profiles, ensuring reliability for sensitive experimental and manufacturing procedures.
  • Meets USP grade requirements for water for injection
  • Low conductivity of 0.06 µS/cm
  • Total organic carbon (TOC) content of 50 ppb
  • Endotoxin levels controlled at 0.03 EU/mL
  • Supplied in a 1100L bag configuration