Learn More
Teknova WFI Quality Water, 20L bag
SDP
Catalog No. NC3252469
Encompass
WFI Quality Water is a high-grade water that meets both EP and USP grade specifications and is 0.1 m sterile filtered at the time of fill. It is ideal for producing cell culture media, reconstitution of biochemical reagents, or as a final-rinse water for critical applications.
Catalog No. NC3252469 Supplier Teknova Supplier No. 20142
WFI Quality Water is a high-grade water that meets both EP and USP grade specifications and is 0.1 m sterile filtered at the time of fill. It is ideal for producing cell culture media, reconstitution of biochemical reagents, or as a final-rinse water for critical applications.