Learn More
MyBioSource WGA ELISA KIT 96 STRP WELL
SDP
Supplier:  MyBioSource MBS931166696STRIPWELLS
Encompass_Preferred
Wheat germ agglutinin, WGA ELISA Kit.Assay Type:Quantitative Sandwich;Detection Range:0.25ng/ml-8ng/ml;Sensitivity:0.1ng/mlMYBIOSOURCE mfg# MBS9311666
Catalog No. NC2199668