Learn More
Biotium Wheat Germ Agglutinin, HRP Conjugate
Catalog No. 501964460
Encompass_Preferred
This product is wheat germ agglutinin (WGA) labeled with horseradish peroxidase (HRP). WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine.WGA conjugates can be used to stain glycoproteins that have these sugar moieties. WGA can be used as a gram stain to label gram bacteria but not gram- bacteria. WGA also binds to the bud scars on budding yeast such as Saccharomyces cerevisiae. WGA HRP conjugate can be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications.
Catalog No. 50-196-4460 Supplier Biotium Supplier No. 29073
This product is wheat germ agglutinin (WGA) labeled with horseradish peroxidase (HRP). WGA is a carbohydrate-binding lectin that has high affinity for sialic acid and N-acetylglucosamine.WGA conjugates can be used to stain glycoproteins that have these sugar moieties. WGA can be used as a gram stain to label gram bacteria but not gram- bacteria. WGA also binds to the bud scars on budding yeast such as Saccharomyces cerevisiae. WGA HRP conjugate can be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications.