Learn More
MTC Bio Weigh Boats, 500/PK
Supplier:  MTC Bio B6501W
Encompass_Preferred
This item has a minimum qty of 5 per supplier requirements.
Weigh Boat, white, 5ml, 35 x 55mm, diamond shaped
Catalog No. 50-187-0996