Learn More
Enzo Life Sciences WH-4-023 (10mg). CAS: 837422-57-8
SDP
Catalog No. 502013447
Encompass_Preferred
Alternative name: 2,6-Dimethylphenyl-N-(2,4-dimethoxyphenyl)-N-[2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]carbamate. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.
Catalog No. 50-201-3447 Supplier Enzo Life Sciences Supplier No. ENZCHM1450010
Alternative name: 2,6-Dimethylphenyl-N-(2,4-dimethoxyphenyl)-N-[2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]-4-pyrimidinyl]carbamate. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -80°C.