Learn More
Abcam WH-4-023, 25MG
Catalog No. 502671533
Encompass_Preferred
MW 568.7 Da, Purity >98%. Potent and selective inihbitor of the tyrosine kinases Lck and Src with IC₅₀ values of 2 nM for Lck and 6 nM for Src.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1533 Supplier Abcam Supplier No. AB28242325MG
MW 568.7 Da, Purity >98%. Potent and selective inihbitor of the tyrosine kinases Lck and Src with IC₅₀ values of 2 nM for Lck and 6 nM for Src.
The product is subject to the following: Abcam Restricted Use Statement