Learn More
Apexbio Technology LLC WH-4-023, 5mg. Cas: 837422-57-8 MFCD: MFCD28138072. Lck/Src inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B80035
Encompass_Preferred
WH-4-023 is a potent and selective Lck and Src inhibitor with IC50 values of 2 and 6 nM, respectively [1]. It is also an inhibitor of IC50 values of 10, 22 and 60 nM for SIK 1, 2 and 3, respectively [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5450