Learn More
Cospheric LLC WH POLYETHY MICROSPHS 1.25G/CC
SDP
Catalog No. NC2590562
Encompass
White polymer microspheres are made with Titanium Dioxide (TiO2) and are safe to use for personal care applications. White PE spheres are extremely opaque, offering maximum hiding power even in a small layer of spheres. Opaque white particles act as a superior opacifying agent that offers advantages in paints and coatings, cosmetics and personal care applications.
Catalog No. NC2590562 Supplier Cospheric LLC Supplier No. WPMS1.25 1020UM 0.1G
White polymer microspheres are made with Titanium Dioxide (TiO2) and are safe to use for personal care applications. White PE spheres are extremely opaque, offering maximum hiding power even in a small layer of spheres. Opaque white particles act as a superior opacifying agent that offers advantages in paints and coatings, cosmetics and personal care applications.