Learn More
Gilson Company Inc No. 30 Whitehouse Sieve Standards, 5/pack
Catalog No. NC1882683
Encompass_Preferred
Whitehouse Sieve Standards for sieves with No. 30 (600µm) opening sizes are supplied with five vials, each with a nominal weight of 2.5g.
Catalog No. NC1882683 Supplier Gilson Company Inc Supplier No. BS410
Whitehouse Sieve Standards for sieves with No. 30 (600µm) opening sizes are supplied with five vials, each with a nominal weight of 2.5g.