Learn More
Cole-Parmer Cole-Parmer large Square Polystyrene Weigh Boats, White, 330 mL, 500/Pk
Catalog No. NC9405259 Shop All Cole-Parmer Products
Encompass_Preferred
General purpose sample holders. Translucent, rubberized polystyrene withstands temperatures to 180°F, flexes for easy pouring. Biologically inert; resistant to dilute acids, aqueous solutions, alcohols, bases. For use with balances, microwave moisture analyzers, hundreds of other sample handling applications. Flat bottom resists tipping. Contoured sides permit no-slip handling.
Catalog No. NC9405259 Supplier Cole-Parmer Supplier No. 0101725
General purpose sample holders. Translucent, rubberized polystyrene withstands temperatures to 180°F, flexes for easy pouring. Biologically inert; resistant to dilute acids, aqueous solutions, alcohols, bases. For use with balances, microwave moisture analyzers, hundreds of other sample handling applications. Flat bottom resists tipping. Contoured sides permit no-slip handling.