Learn More
Description
Includes
Fluorescent powder and control powder, UV light pen, glycerol, transfer pipets, content background, experiment procedures and safety information
Demonstrates how an infection can spread in a population through contact handshaking.
- Simulated infectious agent is detected under a long UV light (black light)
Order Info
Components for 25 students.
Specifications
Specifications
| For Use With (Application) | Demonstrate how an infection can spread in a population through contact handshaking |
| Includes | Fluorescent powder and control powder, uv light pen, glycerol, transfer pipets, content background, experiment procedures and safety information |
| Class Size | 25 Students |
| Product Type | Educational Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

