Learn More
Cole-Parmer Whatman 1440-150 40 Ashless Quantitative Filter Paper, 8um, 15cm; 100/Box
Supplier:  Cole-Parmer 0664715
Encompass
Our Whatman filter papers offer you flexibility in filter choice without sacrificing quality. All filters are packaged 100 per box. Quantitative filter papers are the choice for gravimetric analysis. High-purity hardened filters have a high wet-strength and are chemical-resistant to handle vacuum filtration or acid/alkali solutions.
Catalog No. NC0976043