Learn More
QIAGEN Indicating Desiccant Pack
Catalog No. 09923360
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
09-923-360 1 g
1 options
Catalog No. 09-923-360 Supplier QIAGEN Supplier No. WB100003
Add to Cart
Add to Cart
For contamination-free collection, storage and transport of DNA samples.
  • Desiccant packets (1 g) with indicator to ensure that FTA Cards remain dry during transport or storage.
  • A color change from blue to pink indicates absorption of moisture.
Specifications
For Use With (Application) Collection, storage, processing and shipping FTA™ and FTA™ elute cards
Product Type Dessicant Packet
Quantity 1 g