Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QIAGEN Resealable Multi-Barrier Pouches

Catalog No. 09923359
Change view
Click to view available options
Quantity:
12 x15 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-923-359 12 x15 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-923-359 Supplier QIAGEN Supplier No. WB100024
Add to Cart
Add to Cart

For contamination-free collection, storage and transport of DNA samples.

  • Resealable Multi-Barrier Pouches for transport or storage of QIAcard FTA CloneSaver.
  • Laminated layers maintain sample integrity and security by protecting the card from exposure to gas or liquid contamination.
  • The outer paper surface is suitable for labeling or writing.
  • The Multi-Barrier Pouch has a resealable closure.
  • To optimize sample integrity FTA Cards are stored in a Multi-Barrier Pouch with a desiccant packet.

Specifications

Format Pouch
Isolation Technology Impregnated Specialized Filter Paper Technology
For Use With (Equipment) CloneSaver™ Card
For Use With (Application) Collection, storage, processing and shipping FTA™ and FTA™ elute cards
Product Type Multi-barrier Pouch
Quantity 12 x15 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.