Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QIAGEN QIAcard FTA CloneSaver

Catalog No. 09923347
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-923-347 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 09-923-347 Supplier QIAGEN Supplier No. WB120028
Add to Cart
Add to Cart
This item is not returnable. View return policy

For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. 96 sample areas per card.

CloneSaver Card is designed for the collection, long-term storage and purification of plasmid and BAC DNA from bacterial clones in a 96-well format.

Specifications

Description Designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96-well format.
Format 96-well Plate
Isolation Technology Impregnated Specialized Filter Paper Technology
For Use With (Application) Designed for the collection, long-term storage, and purification of plasmid and BAC DNA from bacterial clones in a 96 well format.
Product Type CloneSaver Card
Quantity 1 Pk.
Sample Volume 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.