Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QIAGEN QIAcard FTA Indicating Mini

Catalog No. 09924593
Change view
Click to view available options
Quantity:
100 Pk.
25 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-924-593 25 Pk.
09-923-343 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-924-593 Supplier QIAGEN Supplier No. WB120356
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For collection, stabilization, processing, transport and archiving of buccal nucleic acid samples. Indicating QIAcard FTA Mini with 2 sample areas per card.

  • QIAcard Indicating FTA Cards include a pink dye that turns white when a colorless sample, e.g. buccal cells or saliva is applied.
  • Stabilization and protection of nucleic acids at room temperature with for spot volumes of 125 μL.
  • Cell lysis on contact, denatures proteins and protects DNA from degradation.

Specifications

Description Collect, stabilize, process, transport, and archive buccal DNA samples with Indicating FTA Micro Cards.
Format Card
Isolation Technology Impregnated Specialized Filter Paper Technology
Sample Type Buccal Cells, Saliva
For Use With (Application) Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification,Molecular biology
No. of Samples 2
Product Type Indicating FTA Mini Card
Quantity 25 Pk.
Sample Volume 125 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.