Learn More
LAB ALLEY Whatman Grade DE81 Ion Exchange Cellulose Chromatography Paper
SDP
Catalog No. NC3452234
Encompass
Thin (0.20mm) paper with diethylaminoethyl (DEAE) cellulose functional groups. Weakly basic anion exchanger for separation of anions nucleotides and amino acids also for enzyme assays and RNA fingerprinting. SIZE: Each sheet is 11.5 cm x 8 cm. Flowrate: 95mm/30 min. (in water).
Catalog No. NC3452234 Supplier LAB ALLEY Supplier No. 057171A
Thin (0.20mm) paper with diethylaminoethyl (DEAE) cellulose functional groups. Weakly basic anion exchanger for separation of anions nucleotides and amino acids also for enzyme assays and RNA fingerprinting. SIZE: Each sheet is 11.5 cm x 8 cm. Flowrate: 95mm/30 min. (in water).