Learn More
Qiagen SPOT CloneSaver Holder
Catalog No. 09800991
Change view
Click to view available options
Rigid Frame
Catalog No.
09-800-991 Rigid Frame
1 options
Catalog No. 09-800-991 Supplier Qiagen Supplier No. WB100034
SPOT CloneSaver holder
SPOT CloneSaver Holder is a rigid frame that allows automated spotting to standard CloneSaver Cards. It keeps the card flat for uniform and precise spotting of biological samples. The 96-well card is easily inserted into the ANSI/SBS-compatible frame, which can then be placed onto a liquid-handling deck just like a multiwell plate.
The SPOT CloneSaver Holder is compatible with standard liquid handlers manufactured by companies such as Beckman Coulter and Tecan Instruments.
Specifications
Product Type CloneSaver Card Holder
Description Keeps card flat for uniform, precise spotting of biological samples
For Use With (Equipment) Compatible w/standard liquid handlers