Learn More
Bioworld Triticum vulgaris (Wheat) Lectin (WGA) - MicroPlate(s)s
Catalog No. 50197533
Encompass
High quality Wheat Germ Agglutinin coated microplates. The WGA coating of these 96-well microplates offers specific, high affinity binding to the carbohydrate fraction of glycoproteins, enzymes and cell membranes,Technical Specifications:Ligand Source:Triticum vulgarisMolecular Weight:36 kDaCarbohydrate Specificity:(GlcNAcβ4)nInhibiting/Eluting Sugar: Chitin hydrolysateor GlcNAc with acidor saltCarbohydrate for Elution: .2M Chitin hydrolysateor GlcNAc with acidor saltBlood Group Specificity:A, B, OForm:96 Well MicroplateIsoelectric (pI):>9Storage: -20°CApplication note: Calcium is required for binding.
Catalog No. 50-197-533 Supplier Bioworld Supplier No. 201400061
High quality Wheat Germ Agglutinin coated microplates. The WGA coating of these 96-well microplates offers specific, high affinity binding to the carbohydrate fraction of glycoproteins, enzymes and cell membranes,Technical Specifications:Ligand Source:Triticum vulgarisMolecular Weight:36 kDaCarbohydrate Specificity:(GlcNAcβ4)nInhibiting/Eluting Sugar: Chitin hydrolysateor GlcNAc with acidor saltCarbohydrate for Elution: .2M Chitin hydrolysateor GlcNAc with acidor saltBlood Group Specificity:A, B, OForm:96 Well MicroplateIsoelectric (pI):>9Storage: -20°CApplication note: Calcium is required for binding.