Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Wheat Germ agglutinin native Protein from wheat germ

Catalog No. 89932821 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-932-821 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-932-821 Supplier Abnova™ Supplier No. P5361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for Enzyme, SDS-PAGE

Specifications

Formulation Lyophilized from 0.05M Tris-HCl, 0.15M NaCl, pH 8.6
Quantity 5 mg
Regulatory Status RUO
Content And Storage Store at -20°C. After reconstitution with deionized water containing 0.9% sodium chloride, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
For Use With (Application) Enzyme Activity, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Purity or Quality Grade ≥94%
Host Species Plants
Product Type Agglutinin (Wheat Germ) Native Protein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.