Learn More
STEMCELL Technologies Wheat Germ Agglutinin WGA i
Supplier: STEMCELL Technologies 1000816
Fluorescent dye conjugates for staining cell membranes of gram-positive bacteria, mammalian cells, and yeast bud scars - 1000 Ug