Learn More
Bioworld WHEAT GERM AGGLUTININWGA)
Catalog No. 50197705
Encompass
WHEAT GERM AGGLUTININ (WGA) SEPAROPORE 4B-MB: Wheat Germ Agglutinin (Lectin) Separopore 4B Wheat germ (Triticum vulgaris) agglutinin (WGA) is a 36,000 molecular weight protein consisting of two identical subunits. WGA contains a group ...
Catalog No. 50-197-705 Supplier Bioworld Supplier No. 201810183
WHEAT GERM AGGLUTININ (WGA) SEPAROPORE 4B-MB: Wheat Germ Agglutinin (Lectin) Separopore 4B Wheat germ (Triticum vulgaris) agglutinin (WGA) is a 36,000 molecular weight protein consisting of two identical subunits. WGA contains a group ...