Learn More
Biomatik Corporation Wheat Gliadin Polyclonal Antibody
Supplier:  Biomatik Corporation CAC07296
Encompass_Preferred
Wheat Gliadin Polyclonal Antibody is a Plant Polyclonal Antibody to Wheat Gliadin. The Wheat Gliadin Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Plant. Immunogen: Wheat Gliadin (Native Protein). Applications: ELISA, WB. Target Synonyms: Wheat Gliadin. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6713