Learn More
Bioworld Wheat Regeneration Medium (MMS2C), 50 L
Catalog No. 50254808
Encompass_Preferred
Wheat Regeneration Medium (MMS2C)Regeneration medium for monocotsContains MS basal salts with vitamins, sucrose and agar to be supplemented with specific growth regulators.Add 0.001 g/L IAA and 0.002 g/L 6-BAP after autoclavingProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-808 Supplier Bioworld Supplier No. 306301224
Wheat Regeneration Medium (MMS2C)Regeneration medium for monocotsContains MS basal salts with vitamins, sucrose and agar to be supplemented with specific growth regulators.Add 0.001 g/L IAA and 0.002 g/L 6-BAP after autoclavingProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested