Learn More
Bioworld Wheat Rooting Medium (MMSOC), 25 L
Catalog No. 50254810
Encompass_Preferred
Wheat Rooting Medium (MMSOC)Rooting medium for monocotsContains MS basal salts with vitamins, sucrose, agar and supplemented with specific growth regulatorsProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tetsted
Catalog No. 50-254-810 Supplier Bioworld Supplier No. 306301233
Wheat Rooting Medium (MMSOC)Rooting medium for monocotsContains MS basal salts with vitamins, sucrose, agar and supplemented with specific growth regulatorsProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tetsted