Learn More
Bioworld Wheat Rooting Medium (MMSOC), 50 L
Catalog No. 50254811
Encompass_Preferred
Wheat Rooting Medium (MMSOC)Rooting medium for monocotsContains MS basal salts with vitamins, sucrose, agar and supplemented with specific growth regulatorsProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tetsted
Catalog No. 50-254-811 Supplier Bioworld Supplier No. 306301234
Add to Cart
Add to Cart
Wheat Rooting Medium (MMSOC)Rooting medium for monocotsContains MS basal salts with vitamins, sucrose, agar and supplemented with specific growth regulatorsProvided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tetsted