Learn More
DWK Life Sciences Wheaton™ 13 mm Unlined Aluminum Seal Flip-Cap
SDP
Catalog No. 12141489 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color (Seal):
Blue
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color (Seal)
12-141-489 Blue
12-141-488 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-141-489 Supplier DWK Life Sciences Supplier No. 224201LS100
Add to Cart
Add to Cart
Open top seals expose stopper for sample retrieval with a syringe
  • Tear-off seal removes completely allowing content to pour from bottle
  • Solid top seals for long-term storage of samples
  • Flip cap seals provide tamper evidence
TRUSTED_SUSTAINABILITY
Specifications
Color (Seal) Blue
Material (Liner) Unlined Seal
Material Aluminum
Size (Metric) Opening 13 mm
Product Type Flip Cap