Learn More
DWK Life Sciences Wheaton™ 22mm Extractor
SDP
Catalog No. 0294051 Shop All DWK Life Sciences Products
Change view
Click to view available options
Screw Thread Bottom:
24-400
Catalog No. Screw Thread Bottom
02-940-51 24-400
1 options
Catalog No. 02-940-51 Supplier DWK Life Sciences Supplier No. 415502
Add to Cart
Add to Cart
For continuous extraction of analytes from a solid into an organic solvent
  • Eliminates need for hooks, springs and clamps common to standard ground joint designs
TRUSTED_SUSTAINABILITY
Specifications
Product Type Soxhlet Extractor
For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
Certifications/Compliance ASTM E 438
Screw Thread Bottom 24-400
Screw Thread Top 20-400