Learn More
DWK LIFE SCIENCES™ Accessories for Stepper™ 411 Repeater Pipette
SDP
Catalog No. 02912747 Shop All DWK Life Sciences Products
Change view
Click to view available options
Includes:
Bench-top stand
Catalog No. Includes
02-912-747 Bench-top stand
1 options
Catalog No. 02-912-747 Supplier DWK LIFE SCIENCES™ Supplier No. W835010
Add to Cart
Add to Cart
Include a bench-top stand designed to hold one Stepper™ pipette, store two color-coded setting knobs, and accommodate one Ecostep syringe adapter
  • Compatible with the Stepper™ 411 Repeater Pipette
  • Bench-top stand holds one Stepper™ pipette securely
  • Stores two color-coded setting knobs
  • Accommodates one Ecostep syringe adapter
TRUSTED_SUSTAINABILITY
Specifications
Material PVC
Type Pipette Stand
For Use With (Equipment) Stepper™ 411 Repeater Pipette
Includes Bench-top stand