Learn More
DWK LIFE SCIENCES™ WHEATON™ Center Disc Tear-Out Aluminum Seals, Unlined
SDP
Catalog No. 11126206 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Natural
Catalog No. Color
11-126-206 Natural
1 options
Catalog No. 11-126-206 Supplier DWK LIFE SCIENCES™ Supplier No. 224189
Add to Cart
Add to Cart
WHEATON™ Center Disc Tear-Out Aluminum Seals, Unlined remove completely allowing content to pour from bottle. Available in a wide range of sizes and colors with good barrier properties and chemical resistance to alcohol and acetone, among others.
  • Tear-out seal removes completely allowing content to pour from bottle
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among others
  • Wide range of styles and sizes available
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Diameter (Metric) Outer 11 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum None
Quantity 1000/Cs.
Closure Size 11 mm
Autoclavable Not Autoclavable
Product Type Solid Top Cap