Learn More
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
GREENER_CHOICESDP
Catalog No. 11126203 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Red
Closure Size:
13 mm
20 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
11-126-203 Blue 20 mm
06-406-15 Natural 20 mm
11-126-204 Green 13 mm
11-126-250 Red 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-126-203 Supplier DWK Life Sciences Supplier No. 22419305
Only null left
Add to Cart
Add to Cart
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among many others
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Aluminum
Quantity 1000/Cs.
Closure Size 20 mm
Product Type Complete Tear-off Cap