Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ ANTIBind High Purity Sealing Mats
SDP

Catalog No. 02911707 Shop All DWK Life Sciences Products
Change view
Click to view available options
No. of Wells:
384
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
02-911-707 384
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-911-707 Supplier DWK Life Sciences Supplier No. W266046

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High purity natural-colored silcone sealing mats with PTFE spray coating.

  • For use with ANTIBind 384- or 96-well plates
  • Packaged in 5 mats/pack
TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Material PTFE-coated Silicone
No. of Wells 384
No. per Pack 5
Product Type Sealing Mat
Plate Compatibility ANTIBind Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.