Learn More
Description
- Black phenolic open top screw cap without septa
- Open top design provides direct syringe access
- Compatible with 24-400 threaded vials and containers
- Supplied as replacement caps for EPA and laboratory vials
- Available in cases of 200 for high-volume use
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| Sterility | Non-sterile |
| For Use With (Equipment) | Bottle |
| Closure Size | 24-400 |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Screw Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
