Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Black Phenolic Screw Cap With Rubber Liner
SDP

Catalog No. 0645031 Shop All DWK Life Sciences Products
Change view
Click to view available options
Neck Finish:
13-425
18-415
20-400
24-400
38-400
38-415
8-425
No. per Case:
1000 Cs.
200 Cs.
500 Cs.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
06-450-31 38-415 200 Cs.
06-450-29 18-415 500 Cs.
06-450-19 13-425 200 Cs.
06-450-18 8-425 1000 Cs.
06-450-23 24-400 500 Cs.
06-450-22 20-400 500 Cs.
06-450-24 38-400 200 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 06-450-31 Supplier DWK Life Sciences Supplier No. 240269
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Black Phenolic Screw Cap With Rubber Liner features a nontoxic white styrene-butadiene rubber liner and is autoclavable for easy sterilization. Available in multiple GPI finish sizes for general laboratory use.

  • Black phenolic screw cap with cemented-in white styrene-butadiene rubber liner
  • Nontoxic liner material suitable for general laboratory use
  • Autoclavable for easy sterilization
  • Available in a range of GPI finish sizes to fit standard screw thread labware
  • Specially formulated phenolic cap material for durability
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 200 Cs.
Sterility Non-sterile
Neck Finish 38-415
For Use With (Equipment) Bottle
Closure Liner 14B Rubber
Autoclavable Autoclavable
Certifications/Compliance ISO 9001
Disposable Reusable
Product Type Bottle Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.