Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Black Phenolic Solid Top Screw Caps
SDP

Catalog No. 03084125 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
15-425
18-400
20-400
24-400
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03-084-125 24-400
03-084-124 20-400
03-084-120 18-400
03-084-118 15-425
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-084-125 Supplier DWK Life Sciences Supplier No. W239300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Providing safe, secure and chemically inert storage. WHEATON™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.

Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.

  • 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
  • Aluminum foil faced pulp board liners can be used with organic solvents
  • Use LDPE lined caps with distilled water, analytical standards and reagents
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Application) Ideal for cell culture work
Material Phenolic Resin
Sterility Non-sterile
Liner Material Styrene Butadiene Rubber
Closure Size 24-400
Autoclavable Autoclavable
Disposable Reusable
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.