Learn More
Description
Capillary Tubes
Specifications
Specifications
| Material | Borosilicate Glass |
| Sterility | Non-sterile |
| Color | Red |
| Volume (Metric) | 100 to 200 μL |
| Type | Capillary Tube |
| Disposable | Single-use |
| For Use With (Equipment) | Socorex Positive Displacement Micropipette |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
