Learn More
DWK Life Sciences Plunger for 5 - 25uL Pipette
SDP
Catalog No. 1370752 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
5 to 25 μL
Catalog No. Volume (Metric)
13-707-52 5 to 25 μL
1 options
Catalog No. 13-707-52 Supplier DWK Life Sciences Supplier No. 851331
Add to Cart
Add to Cart
Pluger for Acura 841 Positive Displacement Micropipette
TRUSTED_SUSTAINABILITY
Specifications
Type Plunger
Volume (Metric) 5 to 25 μL
For Use With (Equipment) Socorex Positive Displacement Micropipette