Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Caps For Liquid Scintillation Vials
SDP

Catalog No. 0645094 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material (Liner):
Linerless
Metal Foil
Polyethylene Disc
Polyseal™
Material:
Polyethylene
Polypropylene
Urea
Closure Size:
15-425
22-400
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Liner) Material Closure Size
06-450-94 Linerless Polyethylene 22-400
06-450-88 Polyethylene Disc Urea 22-400
06-450-91 Metal Foil Urea 15-425
06-450-90 Polyseal™ Urea 22-400
06-450-92 Metal Foil Urea 22-400
06-450-86 Metal Foil Polypropylene 22-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-450-94 Supplier DWK Life Sciences Supplier No. 241317
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Caps For Liquid Scintillation Vials are screw caps designed for use with WHEATON™ liquid scintillation vials. Use as replacement caps or for vials provided without caps. Choose the right size, cap material, and liner for your application. Not autoclavable.

Screw Caps For Liquid Scintillation Vials

  • Screw caps for WHEATON liquid scintillation vials
  • Use as replacement caps or for vials that are provided without caps
  • Choose the right size screw cap for your vial
  • Select cap and liner material for your application
  • Case of 1000 provides an economical alternative
  • Not autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Description Cap Size: 22-400
Material (Liner) Linerless
Color Natural
Material Polyethylene
Closure Material Polyethylene
Closure Type Screw Cap
Quantity 1000/Cs.
Closure Size 22-400
For Use With (Equipment) Liquid Scintillation Vial
Autoclavable Not Autoclavable
Product Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.