Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
SDP

Catalog No. 0645091 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material (Liner):
Linerless
Metal Foil
Polyethylene Disc
Polyseal™
Material:
Polyethylene
Polypropylene
Urea
Closure Size:
15-425
22-400
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Liner) Material Closure Size
06-450-91 Metal Foil Urea 15-425
06-450-88 Polyethylene Disc Urea 22-400
06-450-90 Polyseal™ Urea 22-400
06-450-92 Metal Foil Urea 22-400
06-450-94 Linerless Polyethylene 22-400
06-450-86 Metal Foil Polypropylene 22-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06-450-91 Supplier DWK Life Sciences Supplier No. 241009
Only null left
Add to Cart
Add to Cart

Use as replacement caps or for vials that are provided without caps

Screw Caps For Liquid Scintillation Vials

  • Screw caps for WHEATON liquid scintillation vials
  • Case of 1000 provides an economical alterative
  • Not autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Description Cap Size: 15-425
Material (Liner) Metal Foil
Color White
Material Urea
Closure Size 15-425
For Use With (Equipment) Liquid Scintillation Vial
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.