Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ Clear Oil Sample Bottle
SDP

Catalog No. 02912143 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-912-143 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-912-143 Supplier DWK Life Sciences Supplier No. W216994
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WHEATON™ Clear Oil Sample Bottle is ideal for storage of many kinds of non-hazardous liquids. Made from clear Type III soda-lime glass and supplied with a cork, it offers a reliable, traditional closure solution for oil and liquid sample collection.

  • Clear Type III soda-lime glass construction
  • Suitable for storage of non-hazardous liquids including oils
  • Bottle supplied with cork inserted
  • WHEATON™ brand quality from DWK Life Sciences
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Color Clear
Diameter (English) 1.45 in.
Includes Attached Cork
Material Soda Lime Glass
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 163 mm
Height (English) 6.41 in.
Diameter (Metric) 37 mm
Autoclavable Not Autoclavable
Capacity (English) 4 oz.
Shape Round
Disposable Reusable
Product Type Sample Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.