Learn More
DWK Life Sciences Wheaton™ Clear-Seal Connecting Adapters
SDP
Catalog No. 153216B Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
13-425
Catalog No. Thread Size
15-321-6B 13-425
1 options
Catalog No. 15-321-6B Supplier DWK Life Sciences Supplier No. 139479
Add to Cart
Add to Cart
Enable interfacing between standard semi-micro glassware and micro systems
  • Feature a Clear-Seal™ inner joint on one end and a screw thread for the WHEATON Connection™ on the other end
  • 13-425 thread size comes with a screw cap and an O-ring to seal thermometers, gas inlet tubes, etc
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I,Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
For Use With (Equipment) Micro systems
Includes Screw cap, O-ring
Material Borosilicate Glass
Product Type Connecting Adapter
Standard Taper Joint No. 19/22
Thread Size 13-425