Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences WHEATON™ COMPLETEPAK™ Sterile Components Seals
GREENER_CHOICESDP

Catalog No. p-7221818 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
13 mm
20 mm
Closure Color:
Blue
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size Closure Color
06451248A 13 mm Blue
06451247A 13 mm Red
06451250A 20 mm Blue
06451249A 20 mm Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 06451248A Supplier DWK Life Sciences Supplier No. W988010S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The WHEATON™ COMPLETEPAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Sterile
  • Flip-off Seal

Certifications

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Blue
Material Aluminum
Sterility Sterile
No. per Case 285/Cs.
Product Line Wheaton CompletePAK
Closure Type Sterile Flip Seal
Closure Size 13 mm
For Use With (Equipment) 2 mL Serum Vial
Autoclavable Autoclavable
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Product Type Sterile Seal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.