Description
- Inert contact surface for safe, contamination-free connections
- Easy to assemble without hooks, springs or clamps
- Screw cap configuration creates a grease-free, vacuum-tight seal
- Can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
- Phenolic connectors have a working temperature of up to 180°C
- Autoclavable for repeated use
Specifications
Specifications
| For Use With (Application) | Used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere |
| Material | Borosilicate Glass (33 Low Extractable) |
| Product Type | Connector |
| Thread Size | 24-400, 24-400 |