Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Connection With Black Phenolic Caps
SDP

Catalog No. p-3304079 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
13-425, 20-400
13-425, 22-400
13-425, 24-400
20-400, 24-400
24-400, 24-400
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
02-912-542 13-425, 22-400
02-912-545 24-400, 24-400
02-912-544 20-400, 24-400
02-912-541 13-425, 20-400
02-912-543 13-425, 24-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02-912-542 Supplier DWK Life Sciences Supplier No. W125495
Add to Cart
Add to Cart

The Wheaton Connection™ Screw Thread Connector Safely joins two exterior threaded glass components without hooks, springs or clamps. Screw cap configuration creates a grease-free, vacuum-tight seal.

  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Phenolic connectors have a working temperature of up to 180°C
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
Product Type Connector
Thread Size 13-425, 22-400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.