Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton Coplin staining ja
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S58916EA
Encompass
Slides extend above the opening permitting them to be manipulated without the use of forceps.
Catalog No. 50-290-1823