Learn More
Sigma Aldrich Fine Chemicals Biosciences Wheaton Coplin staining ja
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S55166EA
Encompass
The top flange of the jar and the lower edge of the cover are ground to prevent evaporation.
Catalog No. 50-290-4675