Learn More
DWK Life Sciences Wheaton™ Cryule™ Polypropylene Caps
SDP
Catalog No. 06450189 Shop All DWK Life Sciences Products
Catalog No. 06-450-189 Supplier DWK Life Sciences Supplier No. 242562
Sterile color-coded 10mm diameter caps
  • Use for easy identification of individual samples
  • Fit externally threaded Cryule round-bottom polypropylene vials with screw caps (03-341-18, -18A, B, and C)
Order Info
  • Packed in polybags of 100 caps per pack
  • Assorted case contains 5 bags with 20 each of red, blue, green, yellow, and pink caps