Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Diagnostic Vial Caps
SDP

Catalog No. 12141606 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Style:
22 R3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Style
12-141-606 Blue 22 R3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-141-606 Supplier DWK Life Sciences Supplier No. 300203B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The vial caps are available in multiple color options to support easy identification and organization of IVD kits. Designed for use with compatible diagnostic vials, they help create complete, reliable kit configurations for laboratory workflows.

  • WHEATON™ diagnostic vial caps available in a choice of 2 different pharmaceutical grade materials
  • Polypropylene linerless caps available in various colours for use with lyophilization stoppers/diagnostic vials
  • Urea caps with EPE liners available in various colors for use with lyophilization stopper/diagnostic vials
  • A choice of colored caps to differentiate reagents in kits
  • Bespoke colored caps available, subject to minimum manufacturing quantities
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Linerless
Color Blue
Material Polypropylene
Pre-slit No
Sterility Non-sterile
For Use With (Application) Sealing Screw‐Thread Diagnostic Vials
Style 22 R3
Quantity 1000
For Use With (Equipment) Diagnostic Vials and 20 mm Freeze Dry Stoppers
Fit for Purpose Certified Quality
Autoclavable Not Autoclavable
Product Type Diagnostic cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.