Learn More
Description
- For both fixed- and variable-volume models
- Precision injection molding care for air-tight fit
- Made from high-grade, virgin polypropylene
- Free from trace metals for excellent accuracy and reproducibility
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Sterility | Sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Clean Claims | DNase, RNase, and Pyrogen free |
| Format | Rack |
| Disposable | Single-use |
| For Use With (Equipment) | Socorex Acura Calibra, Socorex Acura |
| Material | Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
